siRNA Details
| Gene Symbol: Olfr742 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146430.2 | NP_666641.2 | 1 | 37 | ||
| GenBank mRNA | BC132583.1 | - | 1 | 37 | 40130860 | |
More Information
Other Species Targeted
Set Membership: |
|

Olfr742
Genomic Map