siRNA Details
| Gene Symbol: Olfr373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146539.2 | NP_666750.2 | 1 | 927 | ||
| GenBank mRNA | BC106979.2 | - | 1 | 925 | ||
| BC106980.1 | - | 1 | 924 | 40042607 | ||
| BC119444.1 | - | 1 | 927 | |||
| BC119445.2 | - | 1 | 927 | 40042612 | ||

Olfr373
Genomic Map