siRNA Details
| Gene Symbol: Olfr373 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146539.2 | NP_666750.2 | 1 | 769 | ||
| GenBank mRNA | BC106979.2 | - | 1 | 767 | ||
| BC106980.1 | - | 1 | 766 | 40042607 | ||
| BC119444.1 | - | 1 | 769 | |||
| BC119445.2 | - | 1 | 769 | 40042612 | ||

Olfr373
Genomic Map