siRNA Details
| Gene Symbol: Olfr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146829.2 | NP_667040.2 | 2 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr27| Gene Symbol: Olfr27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146829.2 | NP_667040.2 | 2 | 88 | ||