siRNA Details
| Gene Symbol: Olfr1250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146965.1 | NP_667176.1 | 1 | 387 | ||
| GenBank mRNA | BC125323.1 | - | 1 | 481 | 40129838 | |
| BC125349.1 | - | 1 | 481 | 40129864 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1250| Gene Symbol: Olfr1250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146965.1 | NP_667176.1 | 1 | 387 | ||
| GenBank mRNA | BC125323.1 | - | 1 | 481 | 40129838 | |
| BC125349.1 | - | 1 | 481 | 40129864 | ||