siRNA Details
| Gene Symbol: Nav3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081035.1 | NP_001074504.1 | 36 | 6529 | ||
| GenBank mRNA | AK122404.1 | - | 25 | 4339 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nav3| Gene Symbol: Nav3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081035.1 | NP_001074504.1 | 36 | 6529 | ||
| GenBank mRNA | AK122404.1 | - | 25 | 4339 | ||