siRNA Details
| Gene Symbol: Scn1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271045.1 | NP_001257974.1 | 3, 4 | 594 | ||
| NM_001271046.2 | NP_001257975.1 | 3, 4 | 461 | |||
| NM_017288.2 | NP_058984.1 | 3, 4 | 675 | |||
| GenBank mRNA | BC094523.1 | - | 3, 4 | 594 | 7316686 | |
| FQ215379.1 | - | 1, 2 | 252 | |||
| FQ216374.1 | - | 3, 4 | 461 | |||
| M91808.1 | - | 3, 4 | 674 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_011322, U85786, BC009652, BC039140, U46681.

Scn1b
Genomic Map