siRNA Details
| Gene Symbol: Samd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013766.2 | NP_001013788.2 | 8 | 1907 | ||
| GenBank mRNA | AK082201.1 | - | 8 | 1908 | ||
| BC150876.1 | - | 8 | 1908 | 9087787 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samd3| Gene Symbol: Samd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013766.2 | NP_001013788.2 | 8 | 1907 | ||
| GenBank mRNA | AK082201.1 | - | 8 | 1908 | ||
| BC150876.1 | - | 8 | 1908 | 9087787 | ||