siRNA Details
| Gene Symbol: Gabrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017289.1 | NP_058985.1 | 8 | 981 | ||
| GenBank mRNA | BC087714.1 | - | 8 | 946 | 7317154 | |
| M35162.1 | - | 8 | 981 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrd| Gene Symbol: Gabrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017289.1 | NP_058985.1 | 8 | 981 | ||
| GenBank mRNA | BC087714.1 | - | 8 | 946 | 7317154 | |
| M35162.1 | - | 8 | 981 | |||