siRNA Details
| Gene Symbol: Rapgefl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080925.1 | NP_001074394.1 | 14 | 2206 | ||
| GenBank mRNA | AK045359.1 | - | 2 | 127 | ||
| BC080279.1 | - | 13 | 1274 | |||
| BC130266.1 | - | 15 | 1482 | 40143028 | ||

Rapgefl1
Genomic Map