siRNA Details
| Gene Symbol: Mroh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033365.2 | NP_001028537.1 | 11 | 1528 | ||
| GenBank mRNA | AK158799.1 | - | 11 | 1529 | ||
| BC150895.1 | - | 10 | 1431 | 9087806 | ||
| BC172046.1 | - | 10 | 1431 | 9054332 | ||

Mroh5
Genomic Map