siRNA Details
| Gene Symbol: D730002M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052788.1 | - | 2 | 1262 | ||
| AK142601.1 | - | 2 | 1460 | |||
| AK164750.1 | - | 2 | 1460 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D730002M21Rik| Gene Symbol: D730002M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052788.1 | - | 2 | 1262 | ||
| AK142601.1 | - | 2 | 1460 | |||
| AK164750.1 | - | 2 | 1460 | |||