siRNA Details
| Gene Symbol: Cntrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012018.2 | NP_036148.2 | 22, 23 | 3625 | ||
| GenBank mRNA | AK020678.1 | - | 2, 3 | 284 | ||
| AK088733.1 | - | 12, 13 | 1988 | |||
| BC079536.1 | - | 10, 11 | 1650 | |||
| BC158010.1 | - | 21, 22 | 3498 | 9007318 | ||
| X79131.1 | - | 14, 15 | 2357 | |||

Cntrl
Genomic Map