siRNA Details
| Gene Symbol: BC035947 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032005.1 | - | 2 | 1222 | ||
| BC034187.1 | - | 3 | 1162 | 4975064 | ||
| BC035948.1 | - | 1 | 459 | |||
| BC138135.1 | - | 2 | 1176 | 8861154 | ||
| BC138136.1 | - | 2 | 1176 | 8861155 | ||

BC035947
Genomic Map