siRNA Details
| Gene Symbol: Chsy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081163.1 | NP_001074632.1 | 3 | 3027 | ||
| GenBank mRNA | BC153826.1 | - | 2 | 1565 | ||
| BC153827.1 | - | 4 | 2499 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chsy1| Gene Symbol: Chsy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081163.1 | NP_001074632.1 | 3 | 3027 | ||
| GenBank mRNA | BC153826.1 | - | 2 | 1565 | ||
| BC153827.1 | - | 4 | 2499 | |||