siRNA Details
| Gene Symbol: Cacna1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017298.1 | NP_058994.1 | 42, 43 | 5516 | ||
| XM_006252586.2 | XP_006252648.1 | 43, 44 | 5587 | |||
| XM_006252590.2 | XP_006252652.1 | 42, 43 | 5506 | |||
| XM_006252594.2 | XP_006252656.1 | 41, 42 | 5446 | |||
| XM_006252595.2 | XP_006252657.1 | 40, 41 | 5401 | |||
| XM_006252597.2 | XP_006252659.1 | 40, 41 | 5401 | |||
| XM_008770990.1 | XP_008769212.1 | 34, 35 | 6449 | |||
| GenBank mRNA | AF370010.1 | - | 6, 7 | 638 | ||
| D38101.1 | - | 42, 43 | 5516 | |||
| D38102.1 | - | 42, 43 | 5458 | |||

Cacna1d
Genomic Map