siRNA Details
| Gene Symbol: Lrrc36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033371.3 | NP_001028543.1 | 2, 3 | 270 | ||
| NM_001170788.1 | NP_001164259.1 | 5, 6 | 593 | |||
| NM_001170789.1 | NP_001164260.1 | 2, 3 | 270 | |||
| GenBank mRNA | AK133042.1 | - | 2, 3 | 270 | ||
| BC150957.1 | - | 2, 3 | 262 | 9087868 | ||
| BC172120.1 | - | 2, 3 | 262 | 9054406 | ||

Lrrc36
Genomic Map