siRNA Details
| Gene Symbol: Zfp651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166644.1 | NP_001160116.1 | 2 | 833 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp651| Gene Symbol: Zfp651 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166644.1 | NP_001160116.1 | 2 | 833 | ||