siRNA Details
| Gene Symbol: Fam26d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081165.1 | NP_001074634.1 | 2 | 2581 | ||
| GenBank mRNA | AK028763.1 | - | 2 | 2575 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam26d| Gene Symbol: Fam26d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081165.1 | NP_001074634.1 | 2 | 2581 | ||
| GenBank mRNA | AK028763.1 | - | 2 | 2575 | ||