siRNA Details
Gene Symbol: Fam26d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081165.1 | NP_001074634.1 | 2 | 1220 | ||
GenBank mRNA | AK028763.1 | - | 2 | 1214 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam26d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081165.1 | NP_001074634.1 | 2 | 1220 | ||
GenBank mRNA | AK028763.1 | - | 2 | 1214 |