siRNA Details
| Gene Symbol: Phf21b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081166.2 | NP_001074635.2 | 13 | 3423 | ||
| GenBank mRNA | AK033659.1 | - | 2 | 1876 | ||
| AK046530.1 | - | 11 | 3678 | |||
| BC044847.1 | - | 1 | 4275 | |||
| BC048859.1 | - | 1 | 4275 | |||
| BC059040.1 | - | 1 | 2243 | |||
| BC067021.1 | - | 13 | 3067 | |||

Phf21b
Genomic Map