siRNA Details
| Gene Symbol: Cdk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033373.2 | NP_001028545.2 | 10 | 1055 | ||
| GenBank mRNA | AK042881.1 | - | 3 | 264 | ||
| AK211120.1 | - | 3 | 200 | |||
| AK217269.1 | - | 4 | 381 | |||
| BC147314.1 | - | 10 | 1055 | 9056364 | ||
| BC147315.1 | - | 10 | 1055 | 9056370 | ||
| BC147758.1 | - | 10 | 1055 | 9087483 | ||
| BC147763.1 | - | 10 | 1055 | 9087488 | ||
| BC172036.1 | - | 10 | 1052 | 9054322 | ||

Cdk15
Genomic Map