siRNA Details
| Gene Symbol: Cdk15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033373.2 | NP_001028545.2 | 14 | 1401 | ||
| GenBank mRNA | AK042881.1 | - | 5 | 421 | ||
| AK181908.1 | - | 4 | 324 | |||
| AK197819.1 | - | 3 | 290 | |||
| AK219417.1 | - | 4 | 323 | |||
| BC147314.1 | - | 14 | 1401 | 9056364 | ||
| BC147315.1 | - | 14 | 1401 | 9056370 | ||
| BC147758.1 | - | 14 | 1401 | 9087483 | ||
| BC147763.1 | - | 14 | 1401 | 9087488 | ||
| BC172036.1 | - | 14 | 1398 | 9054322 | ||

Cdk15
Genomic Map