siRNA Details
| Gene Symbol: Kcnab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017304.2 | NP_059000.1 | 6, 7 | 772 | ||
| XM_008764342.1 | XP_008762564.1 | 6, 7 | 538 | |||
| XM_008764343.1 | XP_008762565.1 | 6, 7 | 791 | |||
| XM_008764344.1 | XP_008762566.1 | 5, 6 | 495 | |||
| XM_008764345.1 | XP_008762567.1 | 4, 5 | 380 | |||
| GenBank mRNA | X76724.1 | - | 6, 7 | 882 | ||

Kcnab2
Genomic Map