siRNA Details
| Gene Symbol: Gm694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033374.3 | NP_001028546.1 | 2 | 867 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm694| Gene Symbol: Gm694 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033374.3 | NP_001028546.1 | 2 | 867 | ||