siRNA Details
| Gene Symbol: C2cd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017985.2 | NP_001017985.2 | 19 | 3623 | ||
| GenBank mRNA | AK047412.1 | - | 13 | 2281 | ||
| BC094430.1 | - | 18 | 3204 | 30643564 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2cd3| Gene Symbol: C2cd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017985.2 | NP_001017985.2 | 19 | 3623 | ||
| GenBank mRNA | AK047412.1 | - | 13 | 2281 | ||
| BC094430.1 | - | 18 | 3204 | 30643564 | ||