siRNA Details
| Gene Symbol: Pcdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105245.1 | NP_001098715.1 | 6 | 8355 | ||
| NM_001105246.1 | NP_001098716.1 | 5 | 8211 | |||
| GenBank mRNA | AK028278.1 | - | 1 | 828 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdh19| Gene Symbol: Pcdh19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105245.1 | NP_001098715.1 | 6 | 8355 | ||
| NM_001105246.1 | NP_001098716.1 | 5 | 8211 | |||
| GenBank mRNA | AK028278.1 | - | 1 | 828 | ||