siRNA Details
| Gene Symbol: Hist1h4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175654.2 | NP_783585.1 | 1 | 31 | ||
| GenBank mRNA | BC119611.2 | - | 1 | 30 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h4d| Gene Symbol: Hist1h4d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175654.2 | NP_783585.1 | 1 | 31 | ||
| GenBank mRNA | BC119611.2 | - | 1 | 30 | ||