siRNA Details
| Gene Symbol: A230046K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033375.2 | NP_001028547.2 | 17, 18 | 1806 | ||
| GenBank mRNA | AK038582.1 | - | 17, 18 | 1764 | ||
| AK043911.1 | - | 17, 18 | 1777 | |||
| AK129269.1 | - | 12, 13 | 1336 | |||
| AK145000.1 | - | 17, 18 | 1788 | |||
| AK166233.1 | - | 17, 18 | 1779 | |||
| BC020314.2 | - | 14, 15 | 1441 | |||
| BC026467.1 | - | 6, 7 | 767 | |||
| BC145215.1 | - | 17, 18 | 2001 | |||

A230046K03Rik
Genomic Map