siRNA Details
Gene Symbol: A230046K03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033375.2 | NP_001028547.2 | 33 | 4524 | ||
GenBank mRNA | AK089097.1 | - | 4 | 1331 | ||
AK129269.1 | - | 28 | 4054 | |||
AK161247.1 | - | 10 | 1972 | |||
BC145215.1 | - | 33 | 4719 |
