siRNA Details
| Gene Symbol: C530047H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049769.1 | - | 4 | 340 | ||
| BC040266.1 | - | 1249 | ||||
| BC094394.1 | - | 3 | 250 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C530047H08Rik| Gene Symbol: C530047H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049769.1 | - | 4 | 340 | ||
| BC040266.1 | - | 1249 | ||||
| BC094394.1 | - | 3 | 250 | |||