siRNA Details
| Gene Symbol: B230216G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242345.1 | NP_001229274.1 | 6 | 990 | ||
| GenBank mRNA | AK043509.1 | - | 4 | 903 | ||
| AK045625.1 | - | 3 | 1084 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B230216G23Rik| Gene Symbol: B230216G23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242345.1 | NP_001229274.1 | 6 | 990 | ||
| GenBank mRNA | AK043509.1 | - | 4 | 903 | ||
| AK045625.1 | - | 3 | 1084 | |||