siRNA Details
Gene Symbol: B230216G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242345.1 | NP_001229274.1 | 6 | 1400 | ||
GenBank mRNA | AK043509.1 | - | 4 | 1313 | ||
AK045625.1 | - | 3 | 1494 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B230216G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242345.1 | NP_001229274.1 | 6 | 1400 | ||
GenBank mRNA | AK043509.1 | - | 4 | 1313 | ||
AK045625.1 | - | 3 | 1494 |