siRNA Details
| Gene Symbol: Cacna2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033382.2 | NP_001028554.2 | 35, 36 | 3238 | ||
| GenBank mRNA | AK137847.1 | - | 9, 10 | 544 | ||
| BC141091.1 | - | 34, 35 | 3094 | 9055703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacna2d4| Gene Symbol: Cacna2d4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033382.2 | NP_001028554.2 | 35, 36 | 3238 | ||
| GenBank mRNA | AK137847.1 | - | 9, 10 | 544 | ||
| BC141091.1 | - | 34, 35 | 3094 | 9055703 | ||