siRNA Details
Gene Symbol: Cacna2d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033382.2 | NP_001028554.2 | 5 | 625 | ||
GenBank mRNA | AK044427.1 | - | 4 | 2190 | ||
BC141091.1 | - | 5 | 556 | 9055703 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cacna2d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033382.2 | NP_001028554.2 | 5 | 625 | ||
GenBank mRNA | AK044427.1 | - | 4 | 2190 | ||
BC141091.1 | - | 5 | 556 | 9055703 |