siRNA Details
| Gene Symbol: Ctss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017320.1 | NP_059016.1 | 6 | 954 | ||
| GenBank mRNA | FQ218804.1 | - | 1029 | |||
| L03201.1 | - | 6 | 954 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctss| Gene Symbol: Ctss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017320.1 | NP_059016.1 | 6 | 954 | ||
| GenBank mRNA | FQ218804.1 | - | 1029 | |||
| L03201.1 | - | 6 | 954 | |||