siRNA Details
Gene Symbol: E030030I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254745.1 | NP_001241674.1 | 1, 2 | 791 | ||
GenBank mRNA | AK053193.1 | - | 1, 2 | 344 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: E030030I06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254745.1 | NP_001241674.1 | 1, 2 | 791 | ||
GenBank mRNA | AK053193.1 | - | 1, 2 | 344 |