siRNA Details
| Gene Symbol: E030030I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254744.1 | NP_001241673.1 | 2 | 1743 | ||
| GenBank mRNA | AK045260.1 | - | 2 | 1293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E030030I06Rik| Gene Symbol: E030030I06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254744.1 | NP_001241673.1 | 2 | 1743 | ||
| GenBank mRNA | AK045260.1 | - | 2 | 1293 | ||