siRNA Details
| Gene Symbol: A630076J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256174.1 | NP_001243103.1 | 3 | 629 | ||
| GenBank mRNA | AK080367.1 | - | 630 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A630076J17Rik| Gene Symbol: A630076J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256174.1 | NP_001243103.1 | 3 | 629 | ||
| GenBank mRNA | AK080367.1 | - | 630 | |||