siRNA Details
| Gene Symbol: Itpkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081175.1 | NP_001074644.1 | 8 | 4005 | ||
| GenBank mRNA | AK039369.1 | - | 7 | 1920 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itpkb| Gene Symbol: Itpkb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081175.1 | NP_001074644.1 | 8 | 4005 | ||
| GenBank mRNA | AK039369.1 | - | 7 | 1920 | ||