siRNA Details
| Gene Symbol: 3222401L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028276.1 | - | 1 | 2480 | ||
| AK045900.1 | - | 1 | 2486 | |||
| AK086343.1 | - | 1 | 2480 | |||
| AK149307.1 | - | 2008 | ||||
| AV220575.2 | - | 1 | 304 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3222401L13Rik| Gene Symbol: 3222401L13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK028276.1 | - | 1 | 2480 | ||
| AK045900.1 | - | 1 | 2486 | |||
| AK086343.1 | - | 1 | 2480 | |||
| AK149307.1 | - | 2008 | ||||
| AV220575.2 | - | 1 | 304 | |||