siRNA Details
| Gene Symbol: Tmem104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033393.2 | NP_001028565.1 | 6 | 574 | ||
| GenBank mRNA | AK040894.1 | - | 6 | 536 | ||
| AK049868.1 | - | 6 | 511 | |||
| AK131112.1 | - | 6 | 551 | |||
| AK155041.1 | - | 6 | 551 | |||
| AK167098.1 | - | 6 | 530 | |||
| AK171314.1 | - | 6 | 551 | |||
| AK171457.1 | - | 6 | 536 | |||
| BC140976.1 | - | 6 | 494 | 9055588 | ||

Tmem104
Genomic Map