siRNA Details
| Gene Symbol: Fam169a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100458.1 | NP_001093928.1 | 13 | 2385 | ||
| NM_001146045.1 | NP_001139517.1 | 13 | 2352 | |||
| GenBank mRNA | AK033380.1 | - | 1 | 48 | ||
| AK045386.1 | - | 1 | 142 | |||
| AK173051.1 | - | 13 | 2385 | |||
| BC084589.1 | - | 13 | 2365 | |||

Fam169a
Genomic Map