siRNA Details
| Gene Symbol: Fam169a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100458.1 | NP_001093928.1 | 13 | 3802 | ||
| NM_001146045.1 | NP_001139517.1 | 13 | 3769 | |||
| GenBank mRNA | AK033380.1 | - | 1 | 1464 | ||
| AK035190.1 | - | 1 | 622 | |||
| AK045386.1 | - | 1 | 1562 | |||
| AK083815.1 | - | 1 | 325 | |||
| AK173051.1 | - | 13 | 3802 | |||
| BC084589.1 | - | 13 | 3783 | |||

Fam169a
Genomic Map