siRNA Details
| Gene Symbol: Tmem88b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033394.3 | NP_001028566.2 | 2 | 1998 | ||
| GenBank mRNA | AK031860.1 | - | 1 | 417 | ||
| AK038855.1 | - | 903 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem88b| Gene Symbol: Tmem88b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033394.3 | NP_001028566.2 | 2 | 1998 | ||
| GenBank mRNA | AK031860.1 | - | 1 | 417 | ||
| AK038855.1 | - | 903 | ||||