siRNA Details
| Gene Symbol: Soga1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164663.1 | NP_001158135.1 | 15 | 7199 | ||
| GenBank mRNA | AK031493.1 | - | 2 | 2558 | ||
| AK034818.1 | - | 1 | 765 | |||
| AK147227.1 | - | 15 | 7190 | |||
| AK173052.1 | - | 9 | 4373 | |||

Soga1
Genomic Map