siRNA Details
| Gene Symbol: A330050F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145192.1 | NP_001138664.1 | 2 | 1776 | ||
| GenBank mRNA | AK031811.1 | - | 2 | 1885 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A330050F15Rik| Gene Symbol: A330050F15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145192.1 | NP_001138664.1 | 2 | 1776 | ||
| GenBank mRNA | AK031811.1 | - | 2 | 1885 | ||