siRNA Details
| Gene Symbol: Fasn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017332.1 | NP_059028.1 | 32 | 5607 | ||
| GenBank mRNA | M76767.1 | - | 32 | 5607 | ||
| X13415.1 | - | 30 | 5405 | |||
| X62888.1 | - | 32 | 5612 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fasn| Gene Symbol: Fasn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017332.1 | NP_059028.1 | 32 | 5607 | ||
| GenBank mRNA | M76767.1 | - | 32 | 5607 | ||
| X13415.1 | - | 30 | 5405 | |||
| X62888.1 | - | 32 | 5612 | |||