siRNA Details
| Gene Symbol: A230056P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043630.1 | - | 5 | 722 | ||
| AK075669.1 | - | 64 | ||||
| AK140288.1 | - | 5 | 722 | |||
| BC147226.1 | - | 3 | 360 | |||
| BC147227.1 | - | 3 | 360 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230056P14Rik| Gene Symbol: A230056P14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043630.1 | - | 5 | 722 | ||
| AK075669.1 | - | 64 | ||||
| AK140288.1 | - | 5 | 722 | |||
| BC147226.1 | - | 3 | 360 | |||
| BC147227.1 | - | 3 | 360 | |||