siRNA Details
| Gene Symbol: Krt26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033397.5 | NP_001028569.2 | 8 | 2319 | ||
| GenBank mRNA | AK028591.1 | - | 8 | 2319 | ||
| AK028682.1 | - | 7 | 3452 | |||
| AK132445.1 | - | 8 | 2323 | |||
| AK162718.1 | - | 8 | 2321 | |||

Krt26
Genomic Map